Invitrogen
Manufacturer Code:PA529347
Catalog # PIPA529347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 168 and 488 of Human Fibulin 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DHRD DRAD FBLN3 FBNL MLVT MTLV S1-5 EGF-containing fibulin-like extracellular matrix protein 1 FIBL-3 extracellular protein S1-5 fibulin-3; EGF containing fibulin-like extracellular matrix protein 1; EGF-containing fibulin-like extracellular matrix protein 1; epidermal growth factor-containing fibulin-like extracellular matrix protein 1; Extracellular protein S1-5; FIBL-3; Fibrillin-like protein; Fibulin-3
Gene Aliases: DHRD; DRAD; EFEMP1; FBLN3; FBNL; FIBL-3; MLVT; MTLV; S1-5; T16
UniProt ID: (Human) Q12805
Entrez Gene ID: (Human) 2202, (Rat) 305604
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.