Invitrogen
Manufacturer Code:PA537163
Catalog # PIPA537163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human Fibulin-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basement-membrane protein 90; BM-90; BM-90/fibulin extracellular matrix glyroprotein; FIBL-1; Fibulin-1; Fibulin1 Fibulin 1 FIBL1 FBLN1 PP213
Gene Aliases: FBLN; FBLN1; FIBL1; PP213
UniProt ID: (Human) P23142, (Mouse) Q08879
Entrez Gene ID: (Human) 2192, (Rat) 315191, (Mouse) 14114
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.