Novus Biologicals
Manufacturer Code:NBP157911
Catalog # NBP157911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to FBLN1(fibulin 1) The peptide sequence was selected from the N terminal of FBLN1 (NP_001987). Peptide sequence CCVKSQETGDLDVGGLQETDKIIEVEEEQEDPYLNDRCRGGGPCKQQCRD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FBLN FIBL1 FIBL-1 fibulin 1 fibulin-1; FIBL-1; Fibulin-1
Gene Aliases: FBLN; FBLN1; FIBL1; PP213
UniProt ID: (Human) P23142
Entrez Gene ID: (Human) 2192
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.