R&D Systems
Manufacturer Code:MAB1918
Catalog # MAB1918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | P1H11 |
Immunogen | Human plasma-derived Fibronectin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Anastellin; CIG; CIG ED-B fibronectin 1 FINC FN FN1 FNZ GFND GFND2 LETS MSF SMDCF; Cold-insoluble globulin; Fibronectin; FN; migration-stimulating factor; Ugl-Y1; Ugl-Y2; Ugl-Y3
Gene Aliases: CIG; ED-B; FINC; FN; FN1; FNZ; GFND; GFND2; LETS; MSF
UniProt ID: (Human) P02751
Entrez Gene ID: (Human) 2335
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.