Invitrogen
Manufacturer Code:PA529578
Catalog # PIPA529578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 396 and 689 of Human Fibronectin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Anastellin; CIG; CIG ED-B FINC FN FNZ GFND GFND2 LETS MSF cold-insoluble globulin fibronectin migration-stimulating factor; Cold-insoluble globulin; Fibronectin; FN; migration-stimulating factor; Ugl-Y1; Ugl-Y2; Ugl-Y3
Gene Aliases: CIG; E330027I09; ED-B; FINC; FN; Fn-1; FN1; FNZ; GFND; GFND2; LETS; MSF
UniProt ID: (Human) P02751, (Mouse) P11276
Entrez Gene ID: (Human) 2335, (Mouse) 14268
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.