R&D Systems
Manufacturer Code:AF5945
Catalog # AF5945
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Fibromodulin/FMOD Asp75-Ile376 Accession # Q06828 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Collagen-binding 59 kDa protein; Collagen-binding 59 kDa protein fibromodulin fibromodulin proteoglycan FM FMOD Keratan sulfate proteoglycan fibromodulin SLRR2E SLRR2EKSPG fibromodulin; Fibromodulin; FM; Keratan sulfate proteoglycan fibromodulin; KSPG fibromodulin
Gene Aliases: FM; FMOD; SLRR2E
UniProt ID: (Human) Q06828
Entrez Gene ID: (Human) 2331
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.