Proteintech
Manufacturer Code:601081IG150UL
Catalog # 50-173-6244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3E9D10 |
Immunogen | Fibromodulin Fusion Protein Ag4030 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Collagen-binding 59 kDa protein; Fibromodulin; fibromodulin FM FMOD KSPG fibromodulin SLRR2E; FM; Keratan sulfate proteoglycan fibromodulin; KSPG fibromodulin
Gene Aliases: FM; FMOD; SLRR2E
UniProt ID: (Human) Q06828
Entrez Gene ID: (Human) 2331
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.