Proteintech
Manufacturer Code:132811AP150UL
Catalog # 50-172-8622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fibromodulin Fusion Protein Ag4030 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collagen-binding 59 kDa protein; Fibromodulin; fibromodulin FM FMOD KSPG fibromodulin SLRR2E; FM; Keratan sulfate proteoglycan fibromodulin; KSPG fibromodulin
Gene Aliases: AI131919; AU041740; FM; FMOD; SLRR2E
UniProt ID: (Human) Q06828, (Mouse) P50608, (Rat) P50609
Entrez Gene ID: (Human) 2331, (Mouse) 14264, (Rat) 64507
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.