R&D Systems
Manufacturer Code:FAB3715V100UG
Catalog # FAB3715V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 427819 |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human Fibroblast Activation Protein alpha/FAP Leu26-Asp760 Accession # Q12884 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 °C as supplied. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: 170 kDa melanoma membrane-bound gelatinase; 170 kDa melanoma membrane-bound gelatinase DKFZp686G13158 DPPIV EC 3.4.21.- FAP FAPA Fibroblast activation protein alpha fibroblast activation protein alpha Integral membrane serine protease seprase vibronectin; Antiplasmin-cleaving enzyme FAP, soluble form; APCE; Dipeptidyl peptidase FAP; FAPalpha; Fibroblast activation protein alpha; fibroblast activation protein, alpha; Gelatine degradation protease FAP; Integral membrane serine protease; Post-proline cleaving enzyme; Prolyl endopeptidase FAP; Seprase; Serine integral membrane protease; SIMP; Surface-expressed protease
Gene Aliases: DPPIV; FAP; FAPA; SIMP
UniProt ID: (Human) Q12884
Entrez Gene ID: (Human) 2191
Molecular Function:
enzyme modulator
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.