Invitrogen
Manufacturer Code:MA515906
Catalog # PIMA515906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4H9 |
Immunogen | Purified recombit fragment of human FGG expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Fibrinogen gamma chain; fibrinogen gamma-chain; fibrinogen, gamma polypeptide; gamma-fibrinogen; PRO2061 fibrinogen gamma polypeptide; testicular tissue protein Li 70
Gene Aliases: 3010002H13Rik; AI256424; FGG; PRO2061
UniProt ID: (Human) P02679, (Mouse) Q8VCM7
Entrez Gene ID: (Human) 2266, (Mouse) 99571
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.