Invitrogen
Manufacturer Code:710386
Catalog # PI710386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 12 HCLC |
Immunogen | Full length peptide of human Fibrinogen |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: FGA Fib2 Fibrinogen A alpha polypeptide chain Fibrinogen alpha chain; Fibrinogen alpha chain; fibrinogen, A alpha polypeptide; Fibrinopeptide A
Gene Aliases: FGA; Fib2
UniProt ID: (Human) P02671
Entrez Gene ID: (Human) 2243
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.