Invitrogen
Manufacturer Code:MA524394
Catalog # PIMA524394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C2 |
Immunogen | FBN2 (NP&lowbar001990.2 2776 a.a. &bsim 2876 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CCA DA9; fibrillin 5; Fibrillin-2; Fibrillin-2 C-terminal peptide
Gene Aliases: CCA; DA9; EOMD; FBN2
UniProt ID: (Human) P35556
Entrez Gene ID: (Human) 2201
Molecular Function: annexin calcium-binding protein calmodulin cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein extracellular matrix structural protein intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.