Invitrogen
Manufacturer Code:MA534973
Catalog # PIMA534973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL0226 |
Immunogen | Recombinant Protein corresonding to Human Fibrillin 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACMICD AI536462 Asprosin B430209H23 ECTOL1 FBN Fbn1 Fbn-1 Fib-1 fibrillin 1 fibrillin 15 fibrillin-1 GPHYSD2 MASS MFLS MFS1 MP340 OCTD putative SGS SSKS tight skin Tsk WMS WMS2; Asprosin; fibrillin 15; Fibrillin-1
Gene Aliases: ACMICD; ECTOL1; FBN; FBN1; GPHYSD2; MASS; MFLS; MFS1; OCTD; SGS; SSKS; WMS; WMS2
UniProt ID: (Human) P35555
Entrez Gene ID: (Human) 2200
Molecular Function: annexin calcium-binding protein calmodulin cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein extracellular matrix structural protein intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.