Invitrogen
Manufacturer Code:PA527358
Catalog # PIPA527358
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 2679 and 2871 of Human Fibrillin 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACMICD FBN GPHYSD2 MASS MFS1 OCTD SGS SSKS WMS WMS2 fibrillin 15 fibrillin-1; Asprosin; fibrillin 15; Fibrillin-1
Gene Aliases: ACMICD; ECTOL1; FBN; FBN1; GPHYSD2; MASS; MFLS; MFS1; OCTD; SGS; SSKS; WMS; WMS2
UniProt ID: (Human) P35555
Entrez Gene ID: (Human) 2200
Molecular Function:
annexin
calcium-binding protein
calmodulin
cell adhesion molecule
extracellular matrix glycoprotein
extracellular matrix protein
extracellular matrix structural protein
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.