Invitrogen
Manufacturer Code:MA514888
Catalog # PIMA514888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | J.99.5 |
Immunogen | Synthetic peptide surrounding Thr298 of human fibrillarin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 34 kDa nucleolar scleroderma antigen; 34 kDa nucleolar scleroderma antigen 34-kD nucleolar scleroderma antigen RNA U3 small nucleolar interacting protein 1 rRNA 2&prime-O-methyltransferase fibrillarin; 34-kD nucleolar scleroderma antigen; Histone-glutamine methyltransferase; Nucleolar protein 1; RNA, U3 small nucleolar interacting protein 1; rRNA 2'-O-methyltransferase fibrillarin; U6 snRNA 2'-O-methyltransferase fibrillarin
Gene Aliases: AL022665; FBL; FIB; FIB1; FLRN; RNU3IP1
UniProt ID: (Human) P22087, (Rat) P22509, (Mouse) P35550
Entrez Gene ID: (Human) 2091, (Rat) 292747, (Mouse) 14113
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.