Invitrogen
Manufacturer Code:MA316771
Catalog # AMSA6771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 38F3 |
Immunogen | Yeast nuclear preparation (S. cerevisiae) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AT20009p; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; FIB FLRN RNU3IP1; Histone-glutamine methyltransferase; LD24618p; LP11571p; Nucleolar protein 1; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; rRNA 2'-O-methyltransferase fibrillarin; U6 snRNA 2'-O-methyltransferase fibrillarin
Gene Aliases: AL022665; CELE_T01C3.7; CG5725; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; FBL; FIB; fib-1; FIB1; FLRN; PANK; RNU3IP1; T01C3.7
UniProt ID: (Human) P22087, (Rat) P22509, (Mouse) P35550, (C. elegans) Q22053
Entrez Gene ID: (Human) 2091, (Rat) 292747, (Mouse) 14113, (Fruit fly) 46234, (C. elegans) 179999
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.