R&D Systems
Manufacturer Code:MAB3207
Catalog # MAB3207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 349316 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Fgr Met1-Phe82 Accession # P09769 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: c-fgr c-src2 c-src-2 proto-oncogene EC 2.7.10 EC 2.7.10.2 FLJ43153 Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog MGC75096 p55c-fgr p55-c-fgr protein p55-Fgr p58c-fgr Proto-oncogene c-Fgr proto-oncogene tyrosine-protein kinase FGR Src2 SRC2c-fgr protooncogene tyrosine-protein kinase Fgr; c-fgr protooncogene; c-src-2 proto-oncogene; feline Gardner-Rasheed sarcoma viral oncogene homolog; Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog; p55-c-fgr protein; p55-Fgr; p58-Fgr; p58c-Fgr; Proto-oncogene c-Fgr; proto-oncogene tyrosine-protein kinase FGR; Tyrosine-protein kinase Fgr; v-fgr feline Gardner-Rasheed sarcoma viral oncogene homolog
Gene Aliases: c-fgr; c-src2; FGR; p55-Fgr; p55c-fgr; p58-Fgr; p58c-fgr; SRC2
UniProt ID: (Human) P09769
Entrez Gene ID: (Human) 2268
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.