Invitrogen
Manufacturer Code:PA569516
Catalog # PIPA569516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human Fggy |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fggy; FGGY carbohydrate kinase domain-containing protein
Gene Aliases: 2310009E04Rik; FGGY; zgc:85818
UniProt ID: (Human) Q96C11, (Mouse) A2AJL3, (Zebrafish) Q6NUW9
Entrez Gene ID: (Rabbit) 100358699, (Human) 55277, (Pig) 100525819, (Dog) 479554, (Bovine) 534011, (Horse) 100067351, (Mouse) 75578, (Rat) 298250, (Guinea pig) 100727187, (Zebrafish) 406567
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.