Invitrogen
Manufacturer Code:MA537893
Catalog # PIMA537893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2433 |
Immunogen | Recombinant protein of human Fetuin A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 59 kDa bone sialic acid-containing protein A2HS Aa2-066 AHS AHSG alpha 2 HS-glycoprotein alpha 2 (fetuin) alpha 2-HS glycoprotein alpha-2-HS-glycoprotein Alpha-2-HS-glycoprotein chain A Alpha-2-HS-glycoprotein chain B Alpha-2-Z-globulin asialofetuin Ba-alpha-2-glycoprotein BSP Countertrypin FETUA fetuin fetuin A FetuinA fetuin-A Glycoprotein PP63 HSGA phosphorylated N-glycoprotein pp63 pp63 PRO2743; Alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; Ba-alpha-2-glycoprotein; fetuin A; Fetuin-A
Gene Aliases: A2HS; AHS; AHSG; FETUA; HSGA; PRO2743
UniProt ID: (Human) P02765
Entrez Gene ID: (Human) 197
Molecular Function: cysteine protease inhibitor enzyme modulator extracellular matrix glycoprotein extracellular matrix protein protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.