Invitrogen
Manufacturer Code:MA524375
Catalog # PIMA524375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 774761 |
Immunogen | Mouse myeloma cell line NS0-derived recombit rat Fetuin A/AHSG Ala19-I352 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 59 kDa bone sialic acid-containing protein; A2HS AHS FETUA HSGA alpha-2-Z-globulin ba-alpha-2-glycoprotein fetuin-A; alpha 2 HS-glycoprotein alpha 2 (fetuin); Alpha-2-HS-glycoprotein; BSP; Fetuin-A; Glycoprotein PP63; phosphorylated N-glycoprotein pp63
Gene Aliases: Aa2-066; Ahsg; Fetua; pp63
UniProt ID: (Rat) P24090
Entrez Gene ID: (Rat) 25373
Molecular Function: cysteine protease inhibitor enzyme modulator extracellular matrix glycoprotein extracellular matrix protein protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.