Invitrogen
Manufacturer Code:PA595214
Catalog # PIPA595214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse Fetuin A recombit protein (Position: A19-I345). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein Countertrypin Fetuin-A Ahsg Fetua; Countertrypin; Fetuin-A
Gene Aliases: Ahsg; Fetua
UniProt ID: (Mouse) P29699
Entrez Gene ID: (Mouse) 11625
Molecular Function:
cysteine protease inhibitor
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.