Invitrogen
Manufacturer Code:PA578744
Catalog # PIPA578744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human Fetuin A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 59 kDa bone sialic acid-containing protein; alpha 2 HS-glycoprotein alpha 2 (fetuin); Alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein Alpha-2-Z-globulin Ba-alpha-2-glycoprotein Fetuin-A Alpha-2-HS-glycoprotein chain A Alpha-2-HS-glycoprotein chain B AHSG FETUA PRO2743; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; Ba-alpha-2-glycoprotein; BSP; fetuin A; Fetuin-A; Glycoprotein PP63; phosphorylated N-glycoprotein pp63
Gene Aliases: A2HS; Aa2-066; AHS; AHSG; FETUA; HSGA; pp63; PRO2743
UniProt ID: (Human) P02765, (Rat) P24090
Entrez Gene ID: (Human) 197, (Rat) 25373
Molecular Function: cysteine protease inhibitor enzyme modulator extracellular matrix glycoprotein extracellular matrix protein protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.