Novus Biologicals
Manufacturer Code:NBP299957JF646
Catalog # NB086469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Fes (P07332-1 Met1-Arg822). |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.2 feline sarcoma oncogene feline sarcoma virus Proto-oncogene c-Fes Proto-oncogene c-Fps proto-oncogene tyrosine-protein kinase Fes/Fps tyrosine-protein kinase Fes/Fps; feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami avian sarcoma (PRCII) viral (v-fps) oncogene homolog; feline sarcoma oncogene; Feline sarcoma/Fujinami avian sarcoma oncogene homolog; Oncogene FES, feline sarcoma virus; p93c-fes; Proto-oncogene c-Fes; Proto-oncogene c-Fps; proto-oncogene tyrosine-protein kinase Fes/Fps; Tyrosine-protein kinase Fes/Fps
Gene Aliases: FES; FPS
UniProt ID: (Human) P07332
Entrez Gene ID: (Human) 2242
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.