Novus Biologicals
Manufacturer Code:NBP275923SS
Catalog # NBP275923SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1308C |
Immunogen | This Ferroportin/SLC40A1 Antibody (1308C) was developed against a synthetic peptide made to an internal portion of human Ferroportin 1 (within residues 250-320). [UniProt# Q9NP59] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Ferroportin-1; Ferroportin-1 FPN1 FPN1IREG1ferroportin 1 HFE4 HFE4ferroportin-1 IREG1 iron regulated gene 1 Iron-regulated transporter 1 member 3 MST079 MSTP079 MTP1 putative ferroportin 1 variant IIIB SLC11A3 SLC11A3iron regulated gene 1 solute carrier family 11 (proton-coupled divalent metal ion transporters) solute carrier family 11 (proton-coupled divalent metal ion transporters) member 3 solute carrier family 40 (iron-regulated transporter) member 1 solute carrier family 40 member 1; iron regulated gene 1; Iron-regulated transporter 1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3; solute carrier family 40 (iron-regulated transporter), member 1; Solute carrier family 40 member 1
Gene Aliases: FPN1; HFE4; IREG1; MST079; MSTP079; MTP1; SLC11A3; SLC40A1
UniProt ID: (Human) Q9NP59
Entrez Gene ID: (Human) 30061
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.