Invitrogen
Manufacturer Code:PA586928
Catalog # PIPA586928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 120-170 of Human FTH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apoferritin; Cell proliferation-inducing gene 15 protein; Ferritin H subunit; Ferritin heavy chain; ferritin heavy chain 1; Ferritin heavy chain Ferritin H subunit Cell proliferation-inducing gene 15 protein Ferritin heavy chain N-terminally processed FTH1 FTHL6 FTH OK/SW-cl.84 PIG15; Ferritin heavy chain, N-terminally processed; Ferritin subunit H; ferritin, heavy polypeptide 1; H-ferritin; placenta immunoregulatory factor; proliferation-inducing protein 15
Gene Aliases: FHC; FTH; FTH1; FTHL6; HFE5; HFt; MFH; OK/SW-cl.84; PIG15; PLIF
UniProt ID: (Human) P02794, (Mouse) P09528, (Rat) P19132
Entrez Gene ID: (Human) 2495, (Mouse) 14319, (Rat) 25319
Molecular Function: storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.