R&D Systems
Manufacturer Code:MAB93541100
Catalog # MAB93541100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 962609 |
Immunogen | Human Ferritin synthetic peptide Accession # P02794 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: apoferritin; apoferritin Cell proliferation-inducing gene 15 protein Ferritin H subunit ferritin heavy chain ferritin heavy polypeptide 1 FHC FTHEC 1.16.3.1 FTHL6MGC104426 PIG15 placenta immunoregulatory factor PLIF proliferation-inducing protein 15; Cell proliferation-inducing gene 15 protein; Ferritin H subunit; Ferritin heavy chain; Ferritin heavy chain, N-terminally processed; ferritin, heavy polypeptide 1; placenta immunoregulatory factor; proliferation-inducing protein 15
Gene Aliases: FHC; FTH; FTH1; FTHL6; HFE5; OK/SW-cl.84; PIG15; PLIF
UniProt ID: (Human) P02794
Entrez Gene ID: (Human) 2495
Molecular Function: storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.