Novus Biologicals
Manufacturer Code:NBP131944
Catalog # NBP131944
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Ferritin Heavy Chain. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apoferritin; apoferritin Cell proliferation-inducing gene 15 protein Ferritin H subunit ferritin heavy chain ferritin heavy polypeptide 1 FHC FTHEC 1.16.3.1 FTHL6MGC104426 PIG15 placenta immunoregulatory factor PLIF proliferation-inducing protein 15; Cell proliferation-inducing gene 15 protein; Ferritin H subunit; Ferritin heavy chain; Ferritin heavy chain, N-terminally processed; ferritin, heavy polypeptide 1; placenta immunoregulatory factor; proliferation-inducing protein 15
Gene Aliases: FHC; FTH; FTH1; FTHL6; HFE5; OK/SW-cl.84; PIG15; PLIF
UniProt ID: (Human) P02794
Entrez Gene ID: (Human) 2495
Molecular Function: storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.