R&D Systems
Manufacturer Code:MAB7229
Catalog # MAB7229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 690318 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Fer Asn93-Ala302 Accession # P16591 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.2 fer (fps/fes related) tyrosine kinase p94-Fer Phosphoprotein NCP94 Proto-oncogene c-Fer TYK3 TYK3phosphoprotein NCP94 Tyrosine kinase 3 tyrosine-protein kinase Fer; Feline encephalitis virus-related kinase FER; fer (fps/fes related) tyrosine kinase; Fujinami poultry sarcoma/Feline sarcoma-related protein Fer; p94-Fer; phosphoprotein NCP94; protein phosphatase 1, regulatory subunit 74; Proto-oncogene c-Fer; Tyrosine kinase 3; Tyrosine-protein kinase Fer
Gene Aliases: FER; p94-Fer; PPP1R74; TYK3
UniProt ID: (Human) P16591
Entrez Gene ID: (Human) 2241
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.