Invitrogen
Manufacturer Code:PA5114296
Catalog # PIPA5114296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal region of Human APBB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adaptor protein FE65a2; adaptor protein FE65a2 amyloid beta (A4) precursor protein-binding family B member 1 amyloid beta (A4) precursor protein-binding family B member 1 (Fe65) amyloid beta A4 precursor protein-binding family B member 1 amyloid beta precursor protein binding family B member 1 Amyloid-beta A4 precursor protein-binding family B member 1 Apbb1 FE65 MGC:9072 Protein Fe65 RIR stat-like protein; amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65); Amyloid-beta A4 precursor protein-binding family B member 1; Protein Fe65; stat-like protein
Gene Aliases: APBB1; FE65; MGC:9072; RIR
UniProt ID: (Human) O00213
Entrez Gene ID: (Human) 322
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.