Invitrogen
Manufacturer Code:165898025
Catalog # 16-589-8025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | MAR-1 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: fc epsilon r1 Fc epsilon receptor Ia Fc epsilon RI alpha-chain Fc fragment of IgE receptor Ia Fc fragment of IgE high affinity I receptor for alpha polypeptide Fc IgE receptor alpha polypeptide Fc receptor IgE high affinity I alpha polypeptide Fce1a Fc-epsilon RI-alpha fcepsilonri FCER1A FcERI Fcr-5 high affinity immunoglobulin epsilon receptor alpha-subunit high affinity immunoglobulin epsilon receptor subunit alpha IgE Fc receptor subunit alpha immunoglobulin E receptor high-affinity of mast cells alpha polypeptide RP11-550P17.3; Fc-epsilon RI-alpha; FcERI; High affinity immunoglobulin epsilon receptor subunit alpha; IgE Fc receptor subunit alpha
Gene Aliases: Fce1a; fcepsilonri; Fcer1a; FcERI; Fcr-5
UniProt ID: (Mouse) P20489
Entrez Gene ID: (Mouse) 14125
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.