Novus Biologicals
Manufacturer Code:NBP143279AF350
Catalog # NP143279A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 43160 |
Immunogen | The immunogen for this antibody was FceRI alpha. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Armenian Hamster Monoclonal Antibody
Protein Aliases: alpha polypeptide Fc fragment of IgE high affinity I receptor for alpha polypeptide Fc-epsilon RI-alpha FcERI high affinity immunoglobulin epsilon receptor alpha-subunit high affinity immunoglobulin epsilon receptor subunit alpha high-affinity of mast cells alpha polypeptide; Fc epsilon receptor Ia; Fc epsilon RI alpha-chain; Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide; Fc IgE receptor, alpha polypeptide; Fc-epsilon RI-alpha; FcERI; high affinity immunoglobulin epsilon receptor alpha-subunit; High affinity immunoglobulin epsilon receptor subunit alpha; IgE Fc receptor subunit alpha; immunoglobulin E receptor, high-affinity, of mast cells, alpha polypeptide
Gene Aliases: FCE1A; FCER1A; FcERI
UniProt ID: (Human) P12319
Entrez Gene ID: (Human) 2205
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.