Abnova
Manufacturer Code:PAB7155
Catalog # 89-132-999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Fbn2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Fbn2.
For Research Use Only
Protein Aliases: BC063774; Fib-2; Fibrillin-2; Fibrillin-2 C-terminal peptide; KIAA4226; mKIAA4226; sne; sy; syndatyly ems
Gene Aliases: BC063774; Fbn-2; Fbn2; Fib-2; mKIAA4226; sne; sy
UniProt ID: (Mouse) Q61555
Entrez Gene ID: (Mouse) 14119
Molecular Function: annexin calcium-binding protein calmodulin cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein extracellular matrix structural protein intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.