Novus Biologicals
Manufacturer Code:NBP229981
Catalog # NBP229981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 497CT15.2.5 |
Immunogen | This FASN antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 942-973 amino acids from the Central region of human FASN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxyacyl-[acyl-carrier-protein] dehydratase; 3-oxoacyl-[acyl-carrier-protein] reductase; 3-oxoacyl-[acyl-carrier-protein] synthase; [Acyl-carrier-protein] S-acetyltransferase; [Acyl-carrier-protein] S-malonyltransferase; Acyl-[acyl-carrier-protein] hydrolase; EC 2.3.1 EC 2.3.1.85 EC 6.3.3.1 FASshort chain dehydrogenase/reductase family 27X member 1 fatty acid synthase MGC14367 MGC15706 OA-519 SDR27X1; Enoyl-[acyl-carrier-protein] reductase; Fatty acid synthase; short chain dehydrogenase/reductase family 27X, member 1; Type I fatty acid synthase
Gene Aliases: FAS; FASN; OA-519; SDR27X1
UniProt ID: (Human) P49327
Entrez Gene ID: (Human) 2194
Molecular Function: acyltransferase dehydrogenase esterase hydrolase ligase methyltransferase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.