Invitrogen
Manufacturer Code:MA511483
Catalog # PIMA511483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | FCN01 (55K-2) |
Immunogen | Fascin purified from HeLa cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 55 kDa actin-bundling protein; 55 kDa actin-bundling protein Singed drosophila homolog-like actin bundling protein fascin singed-like (fascin homolog sea urchin) singed-like protein; Fascin; fascin 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin-bundling protein; fscn1 {ECO:0000250|UniProtKB:Q16658}; p55; singed-like (fascin homolog, sea urchin); Singed-like protein
Gene Aliases: AI663989; FAN1; Fascin; fascin-1; FSCN1; HSN; p55; SNL
UniProt ID: (Human) Q16658, (Rat) P85845, (Mouse) Q61553
Entrez Gene ID: (Human) 6624, (Rat) 683788, (Mouse) 14086
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.