Invitrogen
Manufacturer Code:PA582210
Catalog # PIPA582210
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FSCN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 55 kDa actin-bundling protein; Fascin; fascin 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin-bundling protein; FLJ38511 p55 SNL; fscn1 {ECO:0000250|UniProtKB:Q16658}; p55; singed-like (fascin homolog, sea urchin); Singed-like protein
Gene Aliases: AI663989; FAN1; Fascin; fascin-1; FSCN1; HSN; p55; SNL
UniProt ID: (Human) Q16658, (Rat) P85845, (Mouse) Q61553
Entrez Gene ID: (Human) 6624, (Rat) 683788, (Mouse) 14086
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.