Proteintech
Manufacturer Code:143841AP150UL
Catalog # 50-172-9414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fascin Fusion Protein Ag5816 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 55 kDa actin bundling protein FAN1 Fascin Fascin 1Fascin FSCN1 HSN p55 Singed like protein SNL; 55 kDa actin-bundling protein; Fascin; fascin 1; fascin homolog 1, actin-bundling protein; p55; singed-like (fascin homolog, sea urchin); Singed-like protein
Gene Aliases: AI663989; FAN1; fascin-1; FSCN1; HSN; p55; SNL
UniProt ID: (Human) Q16658, (Mouse) Q61553
Entrez Gene ID: (Human) 6624, (Mouse) 14086
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.