Abnova
Manufacturer Code:H00000356D01P
Catalog # 89-018-013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FASLG (AAH17502.1 1 a.a. ~ 281 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a full-length human FASLG protein.
For Research Use Only
Protein Aliases: ADAM10-processed FasL form; APL; apoptosis (APO-1) antigen ligand 1; Apoptosis antigen ligand; APT1LG1; APTL; CD178; CD95 ligand; CD95-L; CD95L; Fas antigen ligand; Fas ligand; Fas ligand (TNF superfamily, member 6); FASL; FasL ICD; FasL intracellular domain; mutant tumor necrosis factor family member 6; Receptor-binding FasL ectodomain; sFasL; Soluble Fas ligand; SPA; SPPL2A-processed FasL form; TNFSF6; tumor necrosis factor ligand 1A; Tumor necrosis factor ligand superfamily member 6; Tumor necrosis factor ligand superfamily member 6, membrane form; Tumor necrosis factor ligand superfamily member 6, soluble form
Gene Aliases: ALPS1B; APT1LG1; APTL; CD178; CD95-L; CD95L; FASL; FASLG; TNFSF6; TNLG1A
UniProt ID: (Human) P48023
Entrez Gene ID: (Human) 356
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.