Abnova
Manufacturer Code:H00023017A01
Catalog # 89-004-553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | FAIM2 (AAH00051 1 a.a. ~ 316 a.a) full-length recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant FAIM2.
For Research Use Only
Protein Aliases: Fas apoptotic inhibitory molecule 2; KIAA0950; LFG; Neural membrane protein 35; NGP35; NMP35; Protein lifeguard 2; TMBIM2; Transmembrane BAX inhibitor motif-containing protein 2
Gene Aliases: FAIM2; KIAA0950; LFG; LFG2; NGP35; NMP35; TMBIM2
UniProt ID: (Human) Q9BWQ8
Entrez Gene ID: (Human) 23017
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.