R&D Systems
Manufacturer Code:AF126
Catalog # AF126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Fas Ligand/TNFSF6 Pro134-Leu281 Accession # Q53ZZ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAM10-processed FasL form; APL; apoptosis (APO-1) antigen ligand 1; apoptosis (APO-1) antigen ligand 1 Apoptosis antigen ligand APT1LG1CD95L APTL CD178 CD178 antigen CD95L CD95-L Fas antigen ligand Fas ligand Fas ligand (TNF superfamily member 6) FASLCD95 ligand FASLG TNFSF6 TNFSF6FasL tumor necrosis factor (ligand) superfamily member 6 tumor necrosis factor ligand superfamily member 6; Apoptosis antigen ligand; APTL; CD178; CD95 ligand; CD95-L; Fas antigen ligand; Fas ligand; Fas ligand (TNF superfamily, member 6); FasL ICD; FasL intracellular domain; mutant tumor necrosis factor family member 6; Receptor-binding FasL ectodomain; sFasL; Soluble Fas ligand; SPA; SPPL2A-processed FasL form; tumor necrosis factor ligand 1A; Tumor necrosis factor ligand superfamily member 6; Tumor necrosis factor ligand superfamily member 6, membrane form; Tumor necrosis factor ligand superfamily member 6, soluble form
Gene Aliases: ALPS1B; APT1LG1; APTL; CD178; CD95-L; CD95L; FASL; FASLG; TNFSF6; TNLG1A
UniProt ID: (Human) P48023
Entrez Gene ID: (Human) 356
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.