Novus Biologicals
Manufacturer Code:NBP259607PCP
Catalog # NBP259607T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | F13A1/1683 |
Immunogen | Recombinant fragment of human Factor XIIIa protein (aa46-181) (exact sequence is proprietary) |
Conjugate | PerCP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: bA525O21.1 (coagulation factor XIII A1 polypeptide) coagulation factor XIII A chain coagulation factor XIII A1 polypeptide Coagulation factor XIIIa EC 2.3.2.13 F13Acoagulation factor XIII A polypeptide factor XIIIa fibrin stabilizing factor A subunit fibrinoligase FSF A subunit Protein-glutamine gamma-glutamyltransferase A chain TGase Transglutaminase A chain transglutaminase. plasma; bA525O21.1 (coagulation factor XIII, A1 polypeptide); Coagulation factor XIII A chain; coagulation factor XIII, A polypeptide; coagulation factor XIII, A1 polypeptide; Coagulation factor XIIIa; factor XIIIa; fibrin stabilizing factor, A subunit; fibrinoligase; FSF, A subunit; Protein-glutamine gamma-glutamyltransferase A chain; TGase; Transglutaminase A chain; transglutaminase. plasma
Gene Aliases: F13A; F13A1
UniProt ID: (Human) P00488
Entrez Gene ID: (Human) 2162
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.