Novus Biologicals
Manufacturer Code:NBP2413750.025MG
Catalog # NB019684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10-11-5 |
Immunogen | Affinity purified human factor XII. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-factor XIIa part 1; beta-factor XIIa part 1 coagulation factor XII coagulation factor XII (Hageman factor) coagulation factor XIIa heavy chain coagulation factor XIIa light chain EC 3.4.21 EC 3.4.21.38 HAE3 HAEX HAFbeta-factor XIIa part 2 Hageman factor; Beta-factor XIIa part 2; Coagulation factor XII; coagulation factor XII (Hageman factor); Coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; HAF; Hageman factor
Gene Aliases: F12; HAE3; HAEX; HAF
UniProt ID: (Human) P00748
Entrez Gene ID: (Human) 2161
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.