Invitrogen
Manufacturer Code:PA576120
Catalog # PIPA576120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human Factor XII Heavy chain (HC). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-factor XIIa part 1; Beta-factor XIIa part 2; Coagulation factor XII; coagulation factor XII (Hageman factor); Coagulation factor XII Hageman factor HAF Coagulation factor XIIa heavy chain; Coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; factor XII; HAF; Hageman factor
Gene Aliases: F12; FXII; HAE3; HAEX; HAF
UniProt ID: (Human) P00748, (Rat) D3ZTE0, (Mouse) Q80YC5
Entrez Gene ID: (Human) 2161, (Rat) 306761, (Mouse) 58992
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.