Proteintech
Manufacturer Code:125511AP150UL
Catalog # 50-172-8059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Factor XII Fusion Protein Ag3401 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-factor XIIa part 1; Beta-factor XIIa part 2; Coagulation factor XII; coagulation factor XII (Hageman factor); Coagulation factor XII F12 Factor XII HAE3 HAEX HAF Hageman factor; Coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; factor XII; HAF; Hageman factor
Gene Aliases: F12; FXII; HAE3; HAEX; HAF
UniProt ID: (Human) P00748, (Rat) D3ZTE0, (Mouse) Q80YC5
Entrez Gene ID: (Human) 2161, (Rat) 306761, (Mouse) 58992
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.