Invitrogen
Manufacturer Code:PA529118
Catalog # PIPA529118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 167 and 428 of Human Factor X |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activated factor Xa heavy chain; Coagulation factor X; Factor X heavy chain; Factor X light chain; factor Xa; FX FXA Stuart-Prower factor factor Xa prothrombinase; prothrombinase; Stuart factor; Stuart-Prower factor
Gene Aliases: Cf10; F10; FX; FXA
UniProt ID: (Human) P00742, (Mouse) O88947
Entrez Gene ID: (Human) 2159, (Mouse) 14058
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.