Novus Biologicals
Manufacturer Code:NBP290254AF350
Catalog # NB071575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 226 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Factor VIII protein. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AHF; AHF Antihemophilic factor coagulation factor VIII procoagulant component coagulation factor VIIIc DXS1253E F8Ccoagulation factor VIII factor VIII F8B FVIII HEMAF8B Procoagulant component; Antihemophilic factor; Coagulation factor VIII; coagulation factor VIII A1 domain; coagulation factor VIII C2 domain; coagulation factor VIII, procoagulant component; coagulation factor VIIIc; Factor VIII B chain; factor VIII F8B; Factor VIIIa heavy chain, 200 kDa isoform; Factor VIIIa heavy chain, 92 kDa isoform; Factor VIIIa light chain; Procoagulant component
Gene Aliases: AHF; DXS1253E; F8; F8B; F8C; FVIII; HEMA
UniProt ID: (Human) P00451
Entrez Gene ID: (Human) 2157
Molecular Function:
apolipoprotein
cell adhesion molecule
enzyme modulator
extracellular matrix protein
hydrolase
membrane-bound signaling molecule
metalloprotease
oxidase
oxidoreductase
protease
receptor
serine protease
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.