Invitrogen
Manufacturer Code:PA547475
Catalog # PIPA547475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit mouse Coagulation Factor VII Ala42-Leu446 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Coagulation factor VII; Coagulation factor VII Proconvertin Coagulation factor 7 Serum prothrombin conversion accelerator SPCA; Factor VII heavy chain; Factor VII light chain; Serum prothrombin conversion accelerator
Gene Aliases: AI132620; Cf7; F7; FVII
UniProt ID: (Mouse) P70375
Entrez Gene ID: (Mouse) 14068
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.