Invitrogen
Manufacturer Code:PA521346
Catalog # PIPA521346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 209 and 466 of Factor VII (Uniprot IDNo.P08709) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); Eptacog alfa; Factor VII heavy chain; Factor VII light chain; FVII coagulation protein; FVII coagulation protein SPCA coagulation factor VII eptacog alfa proconvertin serum prothrombin conversion accelerator; Proconvertin; Serum prothrombin conversion accelerator; SPCA
Gene Aliases: AI132620; Cf7; F7; FVII; SPCA
UniProt ID: (Mouse) P70375, (Human) P08709
Entrez Gene ID: (Mouse) 14068, (Human) 2155
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.