Invitrogen
Manufacturer Code:PA595520
Catalog # PIPA595520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human Factor V. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activated protein C cofactor; Coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V Activated protein C cofactor Proaccelerin labile factor Coagulation factor V heavy chain Coagulation factor V light chain F5; Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; factor V Leiden; Proaccelerin, labile factor
Gene Aliases: F5; FVL; PCCF; RPRGL1; THPH2
UniProt ID: (Human) P12259
Entrez Gene ID: (Human) 2153
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.