Invitrogen
Manufacturer Code:MA529254
Catalog # PIMA529254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 34 |
Immunogen | Recombit Human Coagulation Factor IX/FIX/F9 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Christmas factor; Coagulation factor IX; Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22 Christmas factor Plasma thromboplastin component PTC; factor 9; factor IX F9; plasma thromboplastic component; Plasma thromboplastin component; PTC
Gene Aliases: F9; F9 p22; FIX; HEMB; P19; PTC; THPH8
UniProt ID: (Human) P00740
Entrez Gene ID: (Human) 2158
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.