Novus Biologicals
Manufacturer Code:NBP289372AF594
Catalog # NB066576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 14 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Factor IX (AAB59620.1 Met1-Thr461). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Christmas factor; Christmas factor coagulation factor IX EC 3.4.21 EC 3.4.21.22 factor 9 FIX HEMB MGC129641 MGC129642 plasma thromboplastic component Plasma thromboplastin component PTC; Coagulation factor IX; Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; factor 9; factor IX F9; plasma thromboplastic component; Plasma thromboplastin component; PTC
Gene Aliases: F9; F9 p22; FIX; HEMB; P19; PTC; THPH8
UniProt ID: (Human) P00740
Entrez Gene ID: (Human) 2158
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.